Spring til Overskrift
Spring til Hovedindhold
Spring til Sidefod
Udvidet søgning
Udvidet søgning
SDS
Certifikater
Producentdata
0
0,00 kr
Order Entry
Log ind
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreindtastning
Ordrestatus
Din konto
Kurv
Denmark
Ordreindtastning
Ordrestatus
Din konto
Kurv
ContactUsLinkComponent
Cryogenic Vials with Internal Thread, 1.8 mL 1 * 500 Items
Katalog # STMC200-0667
Leverandør:
STEMCELL Technologies
Udvid billedet
Cryogenic Vials with Internal Thread, 1.8 mL 1 * 500 Items
Katalog # STMC200-0667
Leverandør:
STEMCELL Technologies
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Leverandør: EPPENDORF
Pasteur pipette
Leverandør: RATIOLAB
VWR®, Transfer Pipettes, High Performance
Leverandør: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Leverandør: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Leverandør: EPPENDORF
VWR®, Serologiske pipetter, Standard type
Leverandør: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Leverandør: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Leverandør: Corning
VWR® Inoculating Loops and Needles, Disposable
Leverandør: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Leverandør: VWR Collection
VWR®, Overførselspipetter, Standard Line
Leverandør: VWR Collection