ArciTect™ crRNA
Custom-designed CRISPR RNA for guide RNA generation in CRISPR-Cas9 genome editing.
ArciTect™ crRNA is a custom CRISPR RNA (crRNA), one of two synthetic RNA components required to make a guide RNA (gRNA) template for CRISPR-Cas9 genome editing. ArciTect™ crRNA contains a user-specified 19 to 21 base sequence complementary to the genomic location to be cleaved, and a linker sequence complementary to ArciTect™ tracrRNA (Catalog #STMC76016), allowing fast and efficient gRNA duplex formation. ArciTect™ crRNA should be designed so that it is directly upstream of a protospacer adjacent motif (PAM) site (NGG). It contains 2'-O-methyl and phosphorothioate modifications at the first two 5' and 3' terminal residues for optimal stability and editing efficiency.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Pakn:20 Nmo
- Beskrivelse:ArciTect™ crRNA
- Cat. No.:STMC76012
- Cellespecifik:Pluripotent stem cells