Multi-Cassette (Urine)
12 parameter urine multi-cassette, AMP/BAR/BZO/COC/MDMA/MET/MOP/MTD/OPI/PCP/
TCA/THC
TCA/THC
Katalog # SURES12MC
Leverandør: SURESCREEN
Gå til:
Product Details & Documents
The Multi-Cassette only requires a small amount of urine to perform the test, with test results available at 5 minutes. This multiline format test comes with a pipette for an easy-to-use process.
- Multi-drug test incorporated
- Suitable for low-volume sample
- Test up to 12 parameters simultaneously
Specifikationer
- Beskrivelse:12 parameter urine multi-cassette
- Seq:02
- Analyser:AMP/BAR/BZO/COC/MDMA/MET/MOP/MTD/OPI/PCP/
TCA/THC