Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Instrument trays

Instrument trays

360×240×20 mm
Katalog # PSKA2034030010
Leverandør:   P.S. KUNSTSTOFWERKE
Instrument trays
Katalog # PSKA2034030010
Leverandør:  P.S. KUNSTSTOFWERKE
Instrument trays
Instrument trays

360×240×20 mm

Price
2.120,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

Melamin, hvid. poleret.

  • God kemisk resistens
  • Varmeresistent op til 80 °C
  • Robuste og glatte overflader muliggør nem rengøring

Lav profil.

Specifikationer
  • B×D×H:
    360×240×20 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.