Sampling Scoop and Lid
Sampling scoop with lid, 100 ml, -, Polystyren
Katalog # SAMP8916A-100
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
Sampling scoops with lid are designed for easy sample handling.
- Scoop comes with clear, snap-on lid
- Avoids the need for additional sample containers
- Flat bottom for easy weighing
- Sterile and non sterile versions available
- Available from stock
- Fast delivery
This range of scoops with lids are the perfect answer to reducing sampling errors. Simply snap on the clear lid and collect samples straight for analysis without the need to transfer to any additional containers. The flat bottom design makes it easy to weigh the sample.
Specifikationer
- Steril:-
- Kapacitet:100 ml
- Beskrivelse:Sampling scoop with lid
- Materiale:Polystyren
- Seq:05