Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dichlormethan ≥98% stabilised, TECHNICAL

Dichlormethan ≥98% stabilised, TECHNICAL

950 L
Catalog # 25629.950
Health
Supplier:   VWR Chemicals
Dichlormethan ≥98% stabilised, TECHNICAL
Catalog # 25629.950
Supplier:  VWR Chemicals
CAS Number:  75-09-2
Dichlormethan ≥98% stabilised, TECHNICAL
Dichlormethan ≥98% stabilised, TECHNICAL

950 L

CAS Number:  75-09-2
Price
Inquire For Price
Log in for availability
1 * 950 L
Product Details & Documents
Synonyms:  Methylenchlorid
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39,8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1,322 g/cm³ (20 °C)
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :stabiliseret med ethanol 0,15 %
  • Storage Temperature:Stue temperatur
  • UN:1593
  • REACH:01-2119480404-41
Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 98 %

Specifications

  • Pakn:
    950 L

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.