Orders LinkContactUsLinkComponent
Robot-spidser til Agilent
Robot-spidser til Agilent
30 µl, +, -, 25,83 mm, Transparent, Agilent™ VPrep® and Bravo®, 384-head, 10 racks of 384 tips
Katalog # BITX63300092
Leverandør:   BIOTIX
Robot-spidser til Agilent
Katalog # BITX63300092
Leverandør:  BIOTIX
Robot-spidser til Agilent
Robot-spidser til Agilent
30 µl, +, -, 25,83 mm, Transparent, Agilent™ VPrep® and Bravo®, 384-head, 10 racks of 384 tips
Price
10.220,00 kr
Log ind for tilgængelighed
1 * 3.840 Items
Product Details & Documents
Biotix robotic tips for Agilent™ VPrep® and Bravo® platforms are engineered to ensure complete compatibility, requiring no changes in programming scripts in the platform's liquid handling software suite.
  • Made of medical-grade virgin PP
  • Tips are BioReady™ certified to be free of RNase, DNase, and Endotoxins (pyrogens)
  • Available as standard or filtered tips
Manufactured to the highest engineering standards for robotic systems, the ultra-straight tips target the centre of microplate wells precisely, delivering exceptional accuracy and precision without risk of jamming.
Specifikationer
  • Pakket:
    10 racks of 384 tips
  • Steril:
    +
  • Volumen:
    30 µl
  • Længde:
    25,83 mm
  • Seq:
    04
  • Spits farge:
    Transparent
  • Til brug med:
    Agilent™ VPrep® and Bravo®, 384-head
  • Filter:
    -