Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Femto Pulse System

Femto Pulse System

Storage solution, 100 ml, For the placement of capillaries between runs, For use with the Femto Pulse and Oligo Pro II system
Catalog # AGLSGP-435-0100
Supplier:   AGILENT
Femto Pulse System
Catalog # AGLSGP-435-0100
Supplier:  AGILENT
Femto Pulse System
Femto Pulse System

Storage solution, 100 ml, For the placement of capillaries between runs, For use with the Femto Pulse and Oligo Pro II system

Price
554,82 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Pulsed-field capillary electrophoresis system for QC of high molecular weight DNA and RNA samples.

  • Large fragment resolution - separate high molecular weight DNA smears and fragments through 165 kb in about 1,5 hours
  • Femtogram level sensitivity - achieve 10 times higher sensitivity for nucleic acid smears and up to 100 times higher for nucleic acid fragments
  • Simple sample preparation - minimise sample degradation with fewer pipetting steps and stable reagents
  • Powerful analysis software - one intuitive software program automates analysis of diverse sample types
  • Conserve precious sample - unparalleled sensitivity allows you to use less precious sample and achieve superior detection
  • Make workflows more efficient - eliminate pulsed-field gel electrophoresis from long-read NGS library preparation and BAC analysis
  • Desktop PC supplied with a comprehensive 3-year warranty through HP

By using a pulsed-field power supply, large molecular weight DNA fragments can be separated and nucleic acids detected into the femtogram range. Compared to other non-denaturing gel-based analytical instruments, the Femto Pulse system easily achieves 10 times higher sensitivity for nucleic acid smears and up to 100 times higher sensitivity for nucleic acid fragments.

The ability to separate high molecular weight DNA up to 165 kb and detect nucleic acids down to 50 fg/µl input concentration make the Femto Pulse system ideal for long-read NGS QC, gDNA, small RNA, or cfDNA analysis from low concentration samples.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications
  • Beskrivelse:
    Storage solution, 100 ml
  • Details:
    For the placement of capillaries between runs, For use with the Femto Pulse and Oligo Pro II system
Accessories

Product Information

  • Catalog #
    Beskrivelse
    Details
    Availability
    Price
  • AGLSP60-20
    96 well trays for buffer and waste locations, 50 per pack
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log in for availability
    1 * 50 Items - 3.196,59 kr

Product Information

  • Catalog #
    Beskrivelse
    Details
    Availability
    Price
  • AGLSP60-22
    Large open waste tray/tub
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log in for availability
    1 * 1 Items - 166,24 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.