Qualitative DNA Kits
dsDNA 930 reagent kit (75 - 20000 bp), 500, Fragment analyser system kit for genotyping and qualitative analysis of PCR amplicons, microsatellites, and SSR's from 75 - 20000 bp, includes reagents necessary for running 500 samples
Katalog # AGLSDNF-930-K0500
Leverandør: AGILENT
Gå til:
Product Details & Documents
DNA fragment analysis for PCR, genotyping, and microsatellite applications.
- Broad concentration range - all kits have a concentration range of 0.5 to 50 ng/µl
- Reliable sizing - six different kits to choose from with a sizing range varying from 35 to 20,000 bp
- Low sample volume - only 2 µl of sample is required, minimising precious sample loss to QC steps
- Quick separations - electrophoresis times as short as 15 minutes
- High resolution - separate fragments under 300 bp differing by only 3 bp
The qualitative kits provide automated and accurate sizing with relative quantification of DNA fragments. Common uses include the analysis of PCR amplicons, simple sequence repeats (SSRs)/microsatellites, and genotyping. There are six different kits to choose from based on the size of the DNA or PCR fragments, ranging from 35 to 20,000 bp. Specially formulated gels, markers, and ladders are designed for each kit enabling reliable sizing over a broad range with varying applications. High resolution kits enable the separation of fragments under 300 bp with a 3 bp difference.
: For research use only. Not for use in diagnostic procedures.
Specifikationer
- Beskrivelse:dsDNA 930 reagent kit (75 - 20000 bp), 500
- Seq:08
- Details:Fragment analyser system kit for genotyping and qualitative analysis of PCR amplicons, microsatellites, and SSR's from 75 - 20000 bp, includes reagents necessary for running 500 samples