Orders LinkContactUsLinkComponent
RNA Kits
RNA Kits
HS RNA kit (15 nt), Fragment analyser system kit for the quantitative and qualitative analysis of total RNA from 200 - 6000 nt at low concentrations, includes reagents necessary for running 1000 samples
Katalog # AGLSDNF-472-1000
Leverandør:   AGILENT
RNA Kits
Katalog # AGLSDNF-472-1000
Leverandør:  AGILENT
RNA Kits
HS RNA kit (15 nt), Fragment analyser system kit for the quantitative and qualitative analysis of total RNA from 200 - 6000 nt at low concentrations, includes reagents necessary for running 1000 samples
Price
23.125,76 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Analysis of mRNA and total RNA quality control, IVT mRNA size, and detection of total RNA degradation.
  • Broad concentration range - The HS RNA kit (15 nt) has a concentration range of 50 to 5000 pg/µl, while the RNA kit (15 nt) covers 5 to 500 ng/µl
  • High and standard sensitivity kits - Minimise sample dilutions by choosing the kit that has the right concentration range for your needs
  • Reliable sizing - Sizing range from 200 to 6000 nt ensures accurate and precise sizing of RNA samples
  • Quality metrics - RNA Quality Number (RQN) quickly scores the integrity of total RNA
  • Low sample volume - Only 2 µl of sample is required, minimising sample loss to QC steps
  • Specific RNA analysis modes - ProSize data analysis software can be set to analyse eukaryotic, prokaryotic, plant, and mRNA samples
The RNA kits are used for the automated analysis of mRNA and total RNA quality control, including IVT mRNA sizing and detection of degradation. Quality RNA is crucial to many applications, including qPCR, NGS, microarray analysis, and Northern blot analysis.
ProSize data analysis software calculates an RNA Quality Number (RQN), providing a quantitative quality metric for downstream applications. Excellent resolution allows users to easily distinguish between RNA degradation and small RNA, increasing the accuracy of the RQN score.
Advarsel: For research use only. Not for use in diagnostic procedures.
Documents
Specifikationer
  • Beskrivelse:
    HS RNA kit (15 nt)
  • Seq:
    04
  • Details:
    Fragment analyser system kit for the quantitative and qualitative analysis of total RNA from 200 - 6000 nt at low concentrations, includes reagents necessary for running 1000 samples