Large Fragment Analysis Kits
HS large fragment 50 kb kit, Fragment analyser system kit for the analysis of large DNA fragments and smears ranging from 75 - 50000 bp, includes reagents necessary for running 500 samples
AGLSDNF-464-0500
: AGILENT
DNA analysis for large smears, fragment sizing, long read NGS library QC and single molecule sequencing.
- Broad concentration range - Each kit has different input concentration ranges specific for DNA fragments and smears
- Reliable sizing - Kits for large sized DNA fragments and long-read NGS libraries help ensure accurate and precise sizing up to 50 kb
- Low sample volume - Only 2 µl of sample required minimising sample loss to QC steps
The large fragment kits for the fragment analyser systems are used for the automated quality and quantity analysis of large DNA fragments, smears, long-read NGS libraries, and single molecule sequencing. Easy analysis of DNA fragment sizing, quality, and concentration expedites decisions and improves workflow.
There are several quality assurance steps in long-read NGS library QC workflow. Important sample QC checkpoints include: starting DNA material for degradation, sheared DNA to ensure correct sizing, and the final long-read NGS library for sizing and quantification.
: For research use only. Not for use in diagnostic procedures.
- Beskrivelse:HS large fragment 50 kb kit
- Seq:03
- Details:Fragment analyser system kit for the analysis of large DNA fragments and smears ranging from 75 - 50000 bp, includes reagents necessary for running 500 samples