Orders LinkContactUsLinkComponent
Vakuumfiltertragt
Vakuumfiltertragt
Glass filter funnel with silicone stopper No. 8, base (300 ml funnel without flask), 300 ml, Borosilikatglas, 47 mm
Catalog # 514-4000
Vakuumfiltertragt
Catalog # 514-4000
Vakuumfiltertragt
Vakuumfiltertragt
Glass filter funnel with silicone stopper No. 8, base (300 ml funnel without flask), 300 ml, Borosilikatglas, 47 mm
Price
3.130,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Borosilikatglas, aluminiumsklemme.
Ideel til vakuumfiltrering af væsker og afgasning af HPLC-opløsningsmidler og den mobile fase. Med inddeling.
Filtreringsareal: 9,6 cm².
* med prop, støttebase, uden kolbe.
Delivery information: Filtration funnel with No. 8 stopper support base (514-4000); filtration unit with side arm support assembly NS 40/35, with flask (516-8921).
Specifications
  • Kapacitet:
    300 ml
  • Beskrivelse:
    Glass filter funnel with silicone stopper No. 8, base (300 ml funnel without flask)
  • Materiale:
    Borosilikatglas
  • Seq:
    02
  • Til filter Ø:
    47 mm
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 516-8922
    Fritted glass support base with sidearm
    Log in for availability
    1 * 1 Items - 4.020,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 516-7567
    Fritted glass support base with No. 8 silicone stopper
    Log in for availability
    1 * 1 Items - 2.040,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 516-8923
    Glass flask, 1 L for membrane filter funnel 47 mm Ø
    Log in for availability
    1 * 1 Items - 2.180,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 516-7566
    Glass flask, 4 L for membrane filter funnel 47 mm Ø
    Log in for availability
    1 * 1 Items - 3.780,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 516-7565
    Glass funnel, 1 L for membrane filter funnel 47 mm Ø
    Log in for availability
    1 * 1 Items - 2.050,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 516-7564
    Glass funnel, 300 ml for membrane filter funnel 47 mm Ø
    Log in for availability
    1 * 1 Items - 1.720,00 kr