[EN]PFIZER-COV19 VACC PLASMA KIT(SERIES) 1 * 1 Each
Catalog # COVVACPLS1152P
[EN]PFIZER-COV19 VACC PLASMA KIT(SERIES) 1 * 1 Each
Catalog # COVVACPLS1152P
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
PCR tubes and caps, Axygen®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Blender tasker
Supplier: Grade Products
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
96 MicroWell™ plader, Nunc™
Supplier: Thermo Fisher Scientific
Microplate stands
Supplier: DIVERSIFIED BIOTECH
Caps for microcentrifuge tubes with thread
Supplier: Simport Scientific
Cryogenic tubes
Supplier: Brand
Kryobokse, 136×136 mm
Supplier: RATIOLAB
Ice pans and buckets, multi-purpose, Corning®
Supplier: Corning
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Nunc™ Internally-Threaded Universal Tubes and Rack System
Supplier: Thermo Fisher Scientific
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
[EN]QUADRIPERM TISSUE CULTURE DISH 1 * 48 Items
Supplier: SARSTEDT
Cryoboxes, PP/PC, B175-series
Supplier: GLW