[EN]HSV GLYCOPROTEIN D PEPTIDE 1MG 1 * 1 mg
ANTIGEN-1094
[EN]HSV GLYCOPROTEIN D PEPTIDE 1MG 1 * 1 mg
ANTIGEN-1094
Frequently Bought Together
Pasteur pipette
: RATIOLAB
96 MicroWell™ plader, Nunc-Immuno™
: Thermo Fisher Scientific
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Deepwell Plates 96-Well, 2000 µl
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Stoppers for tubes
: Greiner Bio-One
VWR®, Stoppers, PE
: VWR Collection
Serologiske pipetter, Stripette™
: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Serologiske pipetter, Standard type
: VWR Collection
Kryobokse, 133×133 mm
: TENAK
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection