VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and stand for 8 pipettes, 0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl
Katalog # 613-7081
Leverandør: VWR Collection
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Katalog # 613-7081
Leverandør: VWR Collection
Gå til:
Product Details & Documents
VWR® Ultra-High Performance pipettes offer superior accuracy and precision, with a proven ergonomic design which are both lightweight and durable.
- Very low plunger forces
- Universal tip capability
- Automatic locking volume adjustment wheel
- Levered tip ejection and colour coded for volume identification
- Easy in-house calibration and maintenance
Specifikationer
- Volumen:0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl
- Beskrivelse:5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and stand for 8 pipettes
- Seq:26
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Shelf clip for 1 single or multichannel pipette, Standard Line, EHP and UHPShelf clip for 1 single or multichannel pipette, Standard Line, EHP and UHP613-7230Log ind for tilgængelighed1 * 1 Items - 114,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Starter Kit incl. 4 UHP pipettes (0,5 - 10, 2 - 20, 20 - 200, 100 - 1000 µl), 4-position stand (613-1589) and 3 racks of tipsStarter Kit incl. 4 UHP pipettes (0,5 - 10, 2 - 20, 20 - 200, 100 - 1000 µl), 4-position stand (613-1589) and 3 racks of tips613-1609Log ind for tilgængelighed1 * 1 Each