Orders LinkContactUsLinkComponent
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and stand for 8 pipettes, 0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl
Katalog # 613-7081
Leverandør:   VWR Collection
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Katalog # 613-7081
Leverandør:  VWR Collection
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and stand for 8 pipettes, 0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl
Price
9.850,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
VWR® Ultra-High Performance pipettes offer superior accuracy and precision, with a proven ergonomic design which are both lightweight and durable.
  • Very low plunger forces
  • Universal tip capability
  • Automatic locking volume adjustment wheel
  • Levered tip ejection and colour coded for volume identification
  • Easy in-house calibration and maintenance
Specifikationer
  • Volumen:
    0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl
  • Beskrivelse:
    5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and stand for 8 pipettes
  • Seq:
    26
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Shelf clip for 1 single or multichannel pipette, Standard Line, EHP and UHP
    Shelf clip for 1 single or multichannel pipette, Standard Line, EHP and UHP
    613-7230
    Log ind for tilgængelighed
    1 * 1 Items - 114,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Starter Kit incl. 4 UHP pipettes (0,5 - 10, 2 - 20, 20 - 200, 100 - 1000 µl), 4-position stand (613-1589) and 3 racks of tips
    Starter Kit incl. 4 UHP pipettes (0,5 - 10, 2 - 20, 20 - 200, 100 - 1000 µl), 4-position stand (613-1589) and 3 racks of tips
    613-1609