Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume

VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume

VWR® UHP, 0,1 - 2 µl, ±40,0 - ±1,5 %, ≤12,0 - ≤0,7 %, rød
Catalog # 613-7063
Supplier:   VWR Collection
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Catalog # 613-7063
Supplier:  VWR Collection
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume

VWR® UHP, 0,1 - 2 µl, ±40,0 - ±1,5 %, ≤12,0 - ≤0,7 %, rød

Price
2.630,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

VWR® Ultra-High Performance pipettes offer superior accuracy and precision, with a proven ergonomic design which are both lightweight and durable.

  • Very low plunger forces
  • Universal tip capability
  • Automatic locking volume adjustment wheel
  • Levered tip ejection and colour coded for volume identification
  • Easy in-house calibration and maintenance
Specifications
  • Farvekode:
    rød
  • Volumen:
    0,1 - 2 µl
  • Beskrivelse:
    VWR® UHP
  • Seq:
    02
  • Nøjagtighed:
    ±40,0 - ±1,5 %
  • Usikkerhed:
    ≤12,0 - ≤0,7 %
Accessories

Product Information

  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-7230
    Shelf clip for 1 single or multichannel pipette, Standard Line, EHP and UHP
    Log in for availability
    1 * 1 Items - 114,00 kr

Product Information

  • Catalog #
    Beskrivelse
    Availability
    Price
  • 613-1609
    Starter Kit incl. 4 UHP pipettes (0,5 - 10, 2 - 20, 20 - 200, 100 - 1000 µl), 4-position stand (613-1589) and 3 racks of tips
    Log in for availability
    1 * 1 Each

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.