Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® PETG/PC Sample Bottles

VWR® PETG/PC Sample Bottles

10 ml, Sterile, 24 mm, 20-415
Katalog # VWRI210-000050
Leverandør:   VWR Collection
VWR® PETG/PC Sample Bottles
Katalog # VWRI210-000050
Leverandør:  VWR Collection
VWR® PETG/PC Sample Bottles
VWR® PETG/PC Sample Bottles

10 ml, Sterile, 24 mm, 20-415

Price
2.510,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.

  • Made from durable, break-resistant 100% virgin PETG material
  • Suitable for frozen storage down to −70°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability

Bottles are arranged in shrink-wrapped trays and polybag packaging to prevent contamination. Bottles 30 ml and above feature volume graduations. Non sterile bottles are autoclavable.

Certifications: US FDA 21CRF, USP Class VI compliant resin.

Accessories information: Spare closures are available for specific application requirements.

Emballering: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.

Specifikationer
  • Cap størrelse:
    20-415
  • Ø:
    24 mm
  • Model:
    Sterile
  • Kapacitet:
    10 ml
  • Seq:
    08
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.