VWR® PETG/PC Sample Bottles
Product Details & Documents
VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic bottles.
- Made from durable, break-resistant 100% virgin PETG material
- Suitable for frozen storage down to −70°C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Leakproof sealing with integrated lip seal feature
- Sterile and non-pyrogenic, SAL 10⁻⁶
- Low extractables
- Transparency and reduced permeability
Bottles are arranged in shrink-wrapped trays and polybag packaging to prevent contamination. Bottles 30 ml and above feature volume graduations. Non sterile bottles are autoclavable.
Certifications: US FDA 21CRF, USP Class VI compliant resin.
Accessories information: Spare closures are available for specific application requirements.
Emballering: Bottles are packed with band, tray arranged, and poly bagged. Spare closures are bulk packed.
Documents
Certifikater
Søg efter partinummerCoC VWR PC Bottles Extractables and Leachable CoC VWR® Media Bottles, Square, PETG coc_VWRI210-000043_238V-52-16J-210723 coc_VWRI210-000044_244V-52-16J-250723 coc_VWRI210-000045_249V-52-16J-270723
Specifikationer
- Cap størrelse:20-415
- Ø:29,8 mm
- Model:Sterile
- Kapacitet:20 ml
- Seq:03
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



