Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
SteriWare Graduated Beakers

SteriWare Graduated Beakers

100 ml, SteriWare beaker
Catalog # SAMP8008P-100
Supplier:   Sampling Systems
SteriWare Graduated Beakers
Catalog # SAMP8008P-100
Supplier:  Sampling Systems
SteriWare Graduated Beakers
SteriWare Graduated Beakers

100 ml, SteriWare beaker

Price
858,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents

The SteriWare range of greduated beakers are ideal for use in the production of drugs, chemicals and cosmetics.

  • Made from food grade, FDA and EC compliant PP
  • Individually bagged
  • Choice of sterile and non sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom

The range of individually bagged graduated beakers from sampling systems are crucial in the production of drugs, chemicals and cosmetics. As they are indivicually bagged they prevent cross contamination and they have graduation marks and a pouring spout to ensure the convnenient handling of liquids.

Specifications
  • Kapacitet:
    100 ml
  • Beskrivelse:
    SteriWare beaker
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.