Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
SteriWare Graduated Beakers

SteriWare Graduated Beakers

250 ml, SteriWare beaker
Katalog # SAMP8008P-250S
Leverandør:   Sampling Systems
SteriWare Graduated Beakers
Katalog # SAMP8008P-250S
Leverandør:  Sampling Systems
SteriWare Graduated Beakers
SteriWare Graduated Beakers

250 ml, SteriWare beaker

Price
1.990,00 kr
Log ind for tilgængelighed
1 * 40 Items
Product Details & Documents

The SteriWare range of greduated beakers are ideal for use in the production of drugs, chemicals and cosmetics.

  • Made from food grade, FDA and EC compliant PP
  • Individually bagged
  • Choice of sterile and non sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom

The range of individually bagged graduated beakers from sampling systems are crucial in the production of drugs, chemicals and cosmetics. As they are indivicually bagged they prevent cross contamination and they have graduation marks and a pouring spout to ensure the convnenient handling of liquids.

Specifikationer
  • Kapacitet:
    250 ml
  • Beskrivelse:
    SteriWare beaker
  • Seq:
    04