Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Iodacetamid 98%

Iodacetamid 98%

Bottle, 5 g
Catalog # APOSOR59962-5G
Toxic
Health
Supplier:   Apollo Scientific
Iodacetamid 98%
Catalog # APOSOR59962-5G
Supplier:  Apollo Scientific
CAS Number:  144-48-9
Iodacetamid 98%

Bottle, 5 g

CAS Number:  144-48-9
Price
kr165.00
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Iodoacetamide
  • CAS Number:144-48-9
  • Formula:ICH₂CONH₂
  • MDL Number:MFCD00008028
  • Molecular Weight:184.96 g/mol
  • Melting Pt:92…95 °C
  • Melting Pt:92 - 95 °C
  • Storage Temperature:Køleskab
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifications