6-Methoxyindol 98%
Bottle, 10 g
APOSOR0581-10G
: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether
- :3189-13-7
- :C₉H₉NO
- :MFCD00022780
- :147.18 g/mol
- :105 °C/0,2mmHg
- :90…94 °C
- :Stue temperatur
- :221-689-6
- Pakning:Bottle
- Pakn:10 g