Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
6-TAMRA

6-TAMRA

Bottle, 100 mg
Catalog # APOSBIB6288-100MG
Irritant
Supplier:   Apollo Scientific
6-TAMRA
Catalog # APOSBIB6288-100MG
Supplier:  Apollo Scientific
CAS Number:  91809-67-5
6-TAMRA

Bottle, 100 mg

CAS Number:  91809-67-5
Price
1.210,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate
  • CAS Number:91809-67-5
  • Formula:C₂₅H₂₂N₂O₅
  • MDL Number:MFCD01073638
  • Molecular Weight:430.45 g/mol
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.