Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
6-TAMRA

6-TAMRA

Bottle, 50 mg
Katalog # APOSBIB6288-50MG
Irriterende
Leverandør:   Apollo Scientific
6-TAMRA
Katalog # APOSBIB6288-50MG
Leverandør:  Apollo Scientific
CAS nummer:  91809-67-5
6-TAMRA

Bottle, 50 mg

CAS nummer:  91809-67-5
Price
769,00 kr
Log ind for tilgængelighed
1 * 50 mg
Product Details & Documents
Synonymer:  6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate
  • CAS nummer:91809-67-5
  • Formel:C₂₅H₂₂N₂O₅
  • MDL nummer:MFCD01073638
  • Molecular Weight:430.45 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    50 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.