Orders LinkContactUsLinkComponent
Brefeldin A
Bottle, 50 mg
  APOSBIB3042-50MG
Brefeldin A
  APOSBIB3042-50MG
 :  20350-15-6
Brefeldin A
Bottle, 50 mg
 :  20350-15-6
1.310,00 kr
1 * 50 mg
BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.
 :  (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
  •  :20350-15-6
  •  :C₁₆H₂₄O₄
  •  :MFCD00083258
  •  :280.4 g/mol
  •  :200…205 °C
  •  :Køleskab
  •  :2811
  •  :6.1,III
  • Pakning:
    Bottle
  • Pakn:
    50 mg
  • Cat. No.:
    APOSBIB3042-50MG