Brefeldin A
Bottle, 50 mg
APOSBIB3042-50MG
BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.
: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
- :20350-15-6
- :C₁₆H₂₄O₄
- :MFCD00083258
- :280.4 g/mol
- :200…205 °C
- :Køleskab
- :2811
- :6.1,III
- Pakning:Bottle
- Pakn:50 mg
- Cat. No.:APOSBIB3042-50MG