Orders LinkContactUsLinkComponent
Arecoliniumbromid
Bottle, 100 g
  APOSOR28886-100G
Arecoliniumbromid
  APOSOR28886-100G
 :  300-08-3
Arecoliniumbromid
Bottle, 100 g
 :  300-08-3
4.230,00 kr
1 * 100 g
 :  Arecoline hydrobromide
  •  :300-08-3
  •  :C₈H₁₄BrNO₂
  •  :MFCD00039041
  •  :236.11 g/mol
  •  :171…176 °C
  •  :Stue temperatur
  •  :206-087-3
  •  :1544
  •  :6.1,III
  • Pakning:
    Bottle
  • Pakn:
    100 g