Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Arecoliniumbromid

Arecoliniumbromid

Bottle, 25 g
Catalog # APOSOR28886-25G
Toxic
Supplier:   Apollo Scientific
Arecoliniumbromid
Catalog # APOSOR28886-25G
Supplier:  Apollo Scientific
CAS Number:  300-08-3
Arecoliniumbromid

Bottle, 25 g

CAS Number:  300-08-3
Price
1.380,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Arecoline hydrobromide
  • CAS Number:300-08-3
  • Formula:C₈H₁₄BrNO₂
  • MDL Number:MFCD00039041
  • Molecular Weight:236.11 g/mol
  • Melting Pt:171…176 °C
  • Storage Temperature:Stue temperatur
  • EINECS:206-087-3
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.