Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Arecoliniumbromid

Arecoliniumbromid

Bottle, 5 g
Katalog # APOSOR28886-5G
Giftig
Leverandør:   Apollo Scientific
Arecoliniumbromid
Katalog # APOSOR28886-5G
Leverandør:  Apollo Scientific
CAS nummer:  300-08-3
Arecoliniumbromid

Bottle, 5 g

CAS nummer:  300-08-3
Price
439,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  Arecoline hydrobromide
  • CAS nummer:300-08-3
  • Formel:C₈H₁₄BrNO₂
  • MDL nummer:MFCD00039041
  • Molecular Weight:236.11 g/mol
  • Smeltende Pt:171…176 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-087-3
  • FN:1544
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.