ThinCert® 96-Well HTS Inserts
Product Details & Documents
96-well membrane plates are designed for high throughput applications such as transport studies, tissue modeling, co-cultures, as well as migration and invasion assays.
- 96-well system for highthroughput applications
- Automation-friendly design
- High permeability and transparency for optimal diffusion rates and optical features
- Optimized fit for the prevention of media wicking
- Excellent membrane flatness for optimal and reproducible cell culture conditions
- Enlarged access ports – convenient access for pipettes, robots and TEER electrodes
- Sterile
The ThinCert 96-well HTS inserts offer a diverse selection of membrane specifications (pore size and density) within a 96 well format. The automation-friendly inserts are highly versatile and can be utilized for various applications such as transport, secretion, and diffusion studies, migration/invasion experiments, co-cultures, cytotoxicity tests, TEER measurements (Trans Epithelial Electric Resistance) and the generation of tissue models (e.g.,endothelia and epithelia).
The two-component system comprises a 96-well porous membrane plate manufactured entirely from polycarbonate (PC) and a corresponding polystyrene (PS) receiver plate. The tissue culture treated PC membrane allows for an ideal exchange of nutrients and substances, creating in vivo-like cultivation conditions to facilitate optimal cell growth, monolayer formation, and tissue differentiation.
The optimised ports ensure a convenient accessibility of the lower (basolateral) compartment with pipettes, automated liquid handling robots as well as electrodes for transepithelial electric resistance (TEER) measurements. The precise, well-centred fit of the membrane plate prevents wicking and provides stable and reproducible assay conditions.
Certifications: All products are certified to be free of RNase, DNase, and human DNA, and are non-pyrogenic and non-cytotoxic.
Bestillingsoplysninger: The 96 well system consist of the 96 well membrane insert plate with lid and a 96-well non-treated receiver plate.
Specifikationer
- Membran:Optimised transparency
- Porestørrelse:0,40 µm
- Beskrivelse:ThinCert® 96 well HTS insert
- Surface treatment:TC
- Pore density:2×10⁷/cm²
- Cat. No.:392-0465
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



