ThinCert® 96-Well HTS Inserts
Product Details & Documents
96-well membrane plates are designed for high throughput applications such as transport studies, tissue modeling, co-cultures, as well as migration and invasion assays.
- 96-well system for highthroughput applications
- Automation-friendly design
- High permeability and transparency for optimal diffusion rates and optical features
- Optimized fit for the prevention of media wicking
- Excellent membrane flatness for optimal and reproducible cell culture conditions
- Enlarged access ports – convenient access for pipettes, robots and TEER electrodes
- Sterile
The ThinCert 96-well HTS inserts offer a diverse selection of membrane specifications (pore size and density) within a 96 well format. The automation-friendly inserts are highly versatile and can be utilized for various applications such as transport, secretion, and diffusion studies, migration/invasion experiments, co-cultures, cytotoxicity tests, TEER measurements (Trans Epithelial Electric Resistance) and the generation of tissue models (e.g.,endothelia and epithelia).
The two-component system comprises a 96-well porous membrane plate manufactured entirely from polycarbonate (PC) and a corresponding polystyrene (PS) receiver plate. The tissue culture treated PC membrane allows for an ideal exchange of nutrients and substances, creating in vivo-like cultivation conditions to facilitate optimal cell growth, monolayer formation, and tissue differentiation.
The optimised ports ensure a convenient accessibility of the lower (basolateral) compartment with pipettes, automated liquid handling robots as well as electrodes for transepithelial electric resistance (TEER) measurements. The precise, well-centred fit of the membrane plate prevents wicking and provides stable and reproducible assay conditions.
: All products are certified to be free of RNase, DNase, and human DNA, and are non-pyrogenic and non-cytotoxic.
: The 96 well system consist of the 96 well membrane insert plate with lid and a 96-well non-treated receiver plate.
Specifikationer
- Beskrivelse:Receiver plates for ThinCert® 96 well HTS insert
- Seq:05
- Surface treatment:TC
- Cat. No.:392-0462
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning