VWR®, Macro Pipette Tips
Macro pipette tips, 1000 - 5000 µl, +, -, 123,1 mm, Clear, Eppendorf, Socorex, Sartorius Biohit, Rack of 50 tips
Supplier: VWR Collection
Gå til:
Product Details & Documents
Improved accuracy over serological pipettes.
- Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Specifikationer
- Pakket:Rack of 50 tips
- Steril:+
- Volumen:1000 - 5000 µl
- Længde:123,1 mm
- Seq:01
- Spidstype:Macro pipette tips
- Spits farge:Clear
- Til brug med:Eppendorf, Socorex, Sartorius Biohit
- Filter:-
- Cat. No.:613-7097