Orders LinkContactUsLinkComponent
Plum iBox
Plum iBox
Plum iBox 2 with 1×200 ml Plum Eyewash pH Neutral 4,9% phosphate and 1×500 ml Plum Eyewash 0.9% sodium chloride
Katalog # PLUM4921
Leverandør:   Plum Safety
Plum iBox
Katalog # PLUM4921
Leverandør:  Plum Safety
Plum iBox
Plum iBox
Plum iBox 2 with 1×200 ml Plum Eyewash pH Neutral 4,9% phosphate and 1×500 ml Plum Eyewash 0.9% sodium chloride
Price
460,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The new Plum iBox is balancing key features in a modern design and is built in a strong and durable material. See-through window lets users keep track of expiry dates immediately.
  • Clearly visible eyewash bottles
  • Quick and efficient overview of expiry dates and product condition
  • Grab from the wall bracket and quickly bring to injured person
  • Spacious design allows access to bottles fast and easily
  • Easily opens – mounted on the wall or standing on the floor
  • Strong and durable material (EPP)
Release Plum iBox from the wall and quickly bring the eyewash to the injured person. Delivered with wall bracket, eyewash plan, and wall mounting set.
Plum iBox 1 iss uitable for dusty environments and small workplaces, with a risk of getting foreign bodies into the eyes. Plum iBox 2 is suitable for workplaces where there is a risk of getting both foreign bodies and/or acids and alkali in the eye. Plum iBox 3 a comprehensive solution for eyewash. Particularly suitable for workplaces where there is a risk of getting both foreign bodies and acids and alkali in the eye.
Certifications: Install the eyewash station less than 20 m or less than 10 seconds from chemical hazard according to EN 15154-4 recommendation.
Specifikationer
  • Beskrivelse:
    Plum iBox 2 with 1×200 ml Plum Eyewash pH Neutral 4,9% phosphate and 1×500 ml Plum Eyewash 0.9% sodium chloride
  • Seq:
    03