Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tannin USP

Tannin USP

125 g
Catalog # SPCMT1013-125GM
Supplier:   Spectrum Chemical
Tannin USP
Catalog # SPCMT1013-125GM
Supplier:  Spectrum Chemical
CAS Number:  1401-55-4
Tannin USP

125 g

CAS Number:  1401-55-4
Price
1.160,00 kr
Log in for availability
1 * 125 g
Product Details & Documents
Synonyms:  Gallotannin, Tannin
  • CAS Number:1401-55-4
  • Formula:C₇₆H₅₂O₄₆
  • MDL Number:MFCD00066397
  • Molecular Weight:1701.2 g/mol
  • Merck Index:12,09221
  • Melting Pt:218…219 °C
  • Density:>1 g/cm³ (20 °C)
  • Flash Pt:199 °C
  • EINECS:215-753-2
Specifications

Specifications

  • Pakn:
    125 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.