Orders LinkContactUsLinkComponent
Masterflex® USB A Serial Adapter Cables, Avantor®
Masterflex® USB A Serial Adapter Cables, Avantor®
Serial Coms Cable: USB-A to PLC (3 wire), 1,8 m
Katalog # MFLX78006-91
Leverandør:   Avantor Fluid Handling
Masterflex® USB A Serial Adapter Cables, Avantor®
Katalog # MFLX78006-91
Leverandør:  Avantor Fluid Handling
Masterflex® USB A Serial Adapter Cables, Avantor®
Serial Coms Cable: USB-A to PLC (3 wire), 1,8 m
Price
836,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Optimize performance of your Masterflex® MasterSense™ or Ismatec® Reglo Touchscreen pump.
  • Connect your drive via USB A for serial control
Not your standard USB cable. Use one of these cables to connect a Masterflex® MasterSense™ and Ismatec® Reglo Touchscreen pumps to a PC or PLC for serial (RS232) communicaiton. Cables features special capability to support serial interface communication. Chose USB A to USB A for communication to computers. The three wire cable is desinged for serial communcation with PLC applications.
Specifikationer
  • Beskrivelse:
    Serial Coms Cable: USB-A to PLC (3 wire), 1,8 m