Masterflex® USB A Serial Adapter Cables, Avantor®
Serial coms cable USB-A - USB-A, 8'
MFLX78006-90
Optimize performance of your Masterflex® MasterSense™ or Ismatec® Reglo Touchscreen pump.
- Connect your drive via USB A for serial control
Not your standard USB cable. Use one of these cables to connect a Masterflex® MasterSense™ and Ismatec® Reglo Touchscreen pumps to a PC or PLC for serial (RS232) communicaiton. Cables features special capability to support serial interface communication. Chose USB A to USB A for communication to computers. The three wire cable is desinged for serial communcation with PLC applications.
- Beskrivelse:Serial coms cable USB-A - USB-A, 8'
- Seq:01