Orders LinkContactUsLinkComponent
GC septa
BTO premium high-temperature centre guide GC septa, 5 mm, 400 °C
  SGEA298787
GC septa
  SGEA298787
GC septa
GC septa
BTO premium high-temperature centre guide GC septa, 5 mm, 400 °C
864,00 kr
1 * 50 Items
  • Ø:
    5 mm
  • Maks. temp.:
    400 °C
  • Beskrivelse:
    BTO premium high-temperature centre guide GC septa
  • Seq:
    01