Orders LinkContactUsLinkComponent
Accessories: Light and control
Accessories: Light and control
Changing polarizer orientation of BL-ZW2 allows parallel and cross-polarization configurations when using a Dino-Lite polarizing microscope, removable polarizer for plain backlight illumination (Model: BL-ZW2 backlight pad)
Katalog # IDCPBL-ZW2
Leverandør:   Dino-Lite / IDCP B.V.
Accessories: Light and control
Katalog # IDCPBL-ZW2
Leverandør:  Dino-Lite / IDCP B.V.
Accessories: Light and control
Accessories: Light and control
Changing polarizer orientation of BL-ZW2 allows parallel and cross-polarization configurations when using a Dino-Lite polarizing microscope, removable polarizer for plain backlight illumination (Model: BL-ZW2 backlight pad)
Price
3.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Changing polarizer orientation of BL-ZW2 allows parallel and cross-polarization configurations when using a Dino-Lite polarizing microscope, removable polarizer for plain backlight illumination (Model: BL-ZW2 backlight pad)