Accessories: Light and control
Changing polarizer orientation of BL-ZW2 allows parallel and cross-polarization configurations when using a Dino-Lite polarizing microscope, removable polarizer for plain backlight illumination (Model: BL-ZW2 backlight pad)
Katalog # IDCPBL-ZW2
Leverandør: Dino-Lite / IDCP B.V.
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Changing polarizer orientation of BL-ZW2 allows parallel and cross-polarization configurations when using a Dino-Lite polarizing microscope, removable polarizer for plain backlight illumination (Model: BL-ZW2 backlight pad)