Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acid red 33

Acid red 33

Glass, amber, 25 g
Katalog # SPCMDC145-25GM
Irriterende
Leverandør:   Spectrum Chemical
Acid red 33
Katalog # SPCMDC145-25GM
Leverandør:  Spectrum Chemical
CAS nummer:  3567-66-6
Acid red 33

Glass, amber, 25 g

CAS nummer:  3567-66-6
Price
1.680,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

D and C Red No. 33 | Non-graded D and C Red No. 33 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonymer:  D&C Red No. 33, Disodium (3Z)-5-amino-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulphonate, Azo Fuchsine
  • CAS nummer:3567-66-6
  • Formel:C₁₆H₁₁N₃Na₂O₇S₂
  • MDL nummer:MFCD00071877
  • Molecular Weight:467.378 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass, amber
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.