Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acid red 33

Acid red 33

Glass, amber, 100 g
Katalog # SPCMDC145-100GM
Irriterende
Leverandør:   Spectrum Chemical
Acid red 33
Katalog # SPCMDC145-100GM
Leverandør:  Spectrum Chemical
CAS nummer:  3567-66-6
Acid red 33

Glass, amber, 100 g

CAS nummer:  3567-66-6
Price
3.740,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

D and C Red No. 33 | Non-graded D and C Red No. 33 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonymer:  D&C Red No. 33, Disodium (3Z)-5-amino-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulphonate, Azo Fuchsine
  • CAS nummer:3567-66-6
  • Formel:C₁₆H₁₁N₃Na₂O₇S₂
  • MDL nummer:MFCD00071877
  • Molecular Weight:467.378 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Glass, amber
  • Pakn:
    100 g

Specifikationer