Orders LinkContactUsLinkComponent
Magnetic Stirring Bars
Magnetic Stirring Bars
25 mm, 8 mm
Katalog # KART0076700
Leverandør:   KARTELL
Magnetic Stirring Bars
Katalog # KART0076700
Leverandør:  KARTELL
Magnetic Stirring Bars
25 mm, 8 mm
Price
305,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Economical stirring bars designed for a wide variety of applications.
  • Stronger mixing action at lower speeds
  • Can be used as an addition to any standard magnetic stir bar range, creating a viable alternative
  • The sizes shown below are nominal, with ±5% tolerance in length and ±10% in diameter
Suitable for foodstuff.
Specifikationer
  • Ø:
    8 mm
  • Længde:
    25 mm
  • Seq:
    02