Indstøbningsringe
gul
Katalog # KART0291906
Leverandør: KARTELL
Gå til:
Product Details & Documents
Embedding rings made from ABS.
- Dimensions: 41×28×11 mm
Available in the colours white, blue, yellow, green and pink.
Specifikationer
- Farve:gul