Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Caps for Erlenmeyer Shaker Flasks

VWR®, Caps for Erlenmeyer Shaker Flasks

Vent cap for 500 ml, 1000 ml
Katalog # 214-0487
   Lav fremstillingspåvirkning
Leverandør:   VWR Collection
VWR®, Caps for Erlenmeyer Shaker Flasks
Katalog # 214-0487
Leverandør:  VWR Collection
   Lav fremstillingspåvirkning
VWR®, Caps for Erlenmeyer Shaker Flasks
VWR®, Caps for Erlenmeyer Shaker Flasks

Vent cap for 500 ml, 1000 ml

Price
557,00 kr
Log ind for tilgængelighed
1 * 25 Items
Specifikationer
  • Beskrivelse:
    Vent cap for 500 ml, 1000 ml
  • Cat. No.:
    214-0487
  • Miljøcertificering:
    ISO 14001/Certificate No.: 04923E00243R0M
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.