Nylon probe tools, Menda
Nylon probe, with point blade, 150 mm, sort
Katalog # 115-3066
Leverandør: DESCO
Gå til:
Product Details & Documents
Ideal for opening plastic cases such as MP3 players, cell phones, laptops and more. Also a great tool for popping out batteries.
- Nylon finish will not mark delicate surfaces
- May be suitable for use in ESD protected areas
- Temperature resistant
Specifikationer
- Farve:sort
- Beskrivelse:Nylon probe, with point blade, 150 mm
- Seq:03