Orders LinkContactUsLinkComponent
Nylon probe tools, Menda
Nylon probe tools, Menda
Nylon probe, with spudger, 140 mm, sort
Katalog # 115-3065
Leverandør:   DESCO
Nylon probe tools, Menda
Katalog # 115-3065
Leverandør:  DESCO
Nylon probe tools, Menda
Nylon probe, with spudger, 140 mm, sort
Price
25,30 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Ideal for opening plastic cases such as MP3 players, cell phones, laptops and more. Also a great tool for popping out batteries.
  • Nylon finish will not mark delicate surfaces
  • May be suitable for use in ESD protected areas
  • Temperature resistant
Specifikationer
  • Farve:
    sort
  • Beskrivelse:
    Nylon probe, with spudger, 140 mm
  • Seq:
    02