Nylon probe tools, Menda
Nylon probe, with fork blade point, 178 mm, sort
115-3064
: DESCO
Ideal for opening plastic cases such as MP3 players, cell phones, laptops and more. Also a great tool for popping out batteries.
- Nylon finish will not mark delicate surfaces
- May be suitable for use in ESD protected areas
- Temperature resistant
- Farve:sort
- Beskrivelse:Nylon probe, with fork blade point, 178 mm
- Seq:01