[EN]FILTER WHEEL 1 * 1 Items
Katalog # MAPD810602
Leverandør: MAPADA
Gå til:
Product Details & Documents
Specifikationer
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein A chromatography
- Antibody type:Primary
- Epitope:C-terminal
- Form:liquid
- Amino acid number:143 to 172
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:FK506 binding protein 14, 22 kDa
- Cat. No.:USBI035663-APC
- Reactivity:Human
- Conjugation:APC (Allophycocyanin)
- Antigen symbol:FKBP14
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- ELISA:Yes
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



