Orders LinkContactUsLinkComponent
Bone curettes, Martini
Bone curettes, Martini, tip diameter 3 / 3,5 mm, 135 mm
  HAMML-HWK-040-01
Bone curettes, Martini
  HAMML-HWK-040-01
Bone curettes, Martini
Bone curettes, Martini, tip diameter 3 / 3,5 mm, 135 mm
960,00 kr
1 * 1 Items
Stainless steel.
  • Double-ended
  • Serrated
  • Beskrivelse:
    Bone curettes, Martini, tip diameter 3 / 3,5 mm
  • Længde:
    135 mm