Orders LinkContactUsLinkComponent
Bone curettes, Martini
Bone curettes, Martini
Bone curettes, Martini, tip diameter 4 / 4,5 mm, 135 mm
Katalog # HAMML-HWK-042-03
Leverandør:   HAMMACHER KARL
Bone curettes, Martini
Katalog # HAMML-HWK-042-03
Leverandør:  HAMMACHER KARL
Bone curettes, Martini
Bone curettes, Martini, tip diameter 4 / 4,5 mm, 135 mm
Price
960,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel.
  • Double-ended
  • Serrated
Specifikationer
  • Beskrivelse:
    Bone curettes, Martini, tip diameter 4 / 4,5 mm
  • Længde:
    135 mm