Screw Caps for Bottles
Push and turn closure, without insert, 28 mm, hvid, HDPE
Katalog # RIXI8-2205-28-W
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Specifikationer
- Gevind:28 mm
- Farve:hvid
- Beskrivelse:Push and turn closure, without insert
- Materiale:HDPE
- Seq:48