Orders LinkContactUsLinkComponent
Cleanroom Overalls, CleanMax®
Clean manufactured coverall, hvid, 5XL
  114-9587
Cleanroom Overalls, CleanMax®
  114-9587
Cleanroom Overalls, CleanMax®
Cleanroom Overalls, CleanMax®
Clean manufactured coverall, hvid, 5XL
3.850,00 kr
1 * 25 Items
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
 : Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
  • Tøj størrelse:
    5XL
  • Farve:
    hvid
  • Beskrivelse:
    Clean manufactured coverall
  • Seq:
    07
  • Cat. No.:
    114-9587
  • Renrum klasse:
    100/ISO 5
  • Outfit applikationer:
    Cleanroom
  • Lukkestørrelse:
    Med lynlås